Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108138.1 | NP_001101608.1 | 6 | 1011 | ||
XM_006242937.2 | XP_006242999.1 | 6 | 993 | |||
GenBank mRNA | BC168871.1 | - | 6 | 846 |