Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLK8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281431.1 | NP_001268360.1 | 5 | 785 | ||
NM_007196.3 | NP_009127.1 | 6 | 945 | |||
NM_144505.2 | NP_653088.1 | 6 | 1080 | |||
NM_144506.2 | NP_653089.1 | 4 | 522 | |||
NM_144507.2 | NP_653090.1 | 3 | 388 | |||
NR_104008.1 | 5 | 682 | ||||
GenBank mRNA | AA320217.1 | - | 2 | 197 | ||
AB008390.2 | - | 6 | 820 | |||
AB008927.1 | - | 5 | 922 | |||
AB009849.1 | - | 6 | 800 | |||
AF095742.1 | - | 6 | 945 | |||
AY359036.1 | - | 6 | 931 | |||
BC040887.1 | - | 7 | 1256 | 5742016 | ||
BM845575.1 | - | 4 | 488 |