Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007210.3 | NP_009141.2 | 4 | 862 | ||
XM_005268607.1 | XP_005268664.1 | 4 | 1121 | |||
XM_006719214.2 | XP_006719277.1 | 3 | 696 | |||
XM_011537819.2 | XP_011536121.1 | 5 | 1077 | |||
XM_011537822.2 | XP_011536124.1 | 4 | 798 | |||
XM_017018744.1 | XP_016874233.1 | 4 | 935 | |||
XM_017018745.1 | XP_016874234.1 | 5 | 920 | |||
GenBank mRNA | AB265820.1 | - | 3 | 747 | ||
AK074658.1 | - | 3 | 760 | |||
AK296852.1 | - | 5 | 768 | |||
BC035822.1 | - | 4 | 865 | 5552059 | ||
BC114505.1 | - | 2 | 541 | 40006926 | ||
Y08565.1 | - | 2 | 541 |