Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109148.1 | NP_001102618.1 | 3 | 296 | ||
XM_006229400.1 | XP_006229462.1 | 4 | 251 |