Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD1564379 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126295.1 | NP_001119767.1 | 2 | 1700 | ||
XM_006233690.2 | XP_006233752.1 | 2 | 1772 | |||
XM_006233691.2 | XP_006233753.1 | 2 | 1700 | |||
XM_006233692.1 | XP_006233754.1 | 2 | 1526 | |||
GenBank mRNA | BC161868.1 | - | 2 | 1700 | 7108986 |