Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008764441.1 | XP_008762663.1 | 7 | 826 | ||
XM_008764442.1 | XP_008762664.1 | 7 | 826 | |||
XM_008764443.1 | XP_008762665.1 | 7 | 826 | |||
XM_008764444.1 | XP_008762666.1 | 7 | 772 | |||
XM_008764445.1 | XP_008762667.1 | 7 | 826 | |||
XM_008764446.1 | XP_008762668.1 | 7 | 826 | |||
XM_008764447.1 | XP_008762669.1 | 7 | 826 | |||
XM_008764448.1 | XP_008762670.1 | 7 | 826 | |||
XM_008764449.1 | XP_008762671.1 | 7 | 826 | |||
XM_008764450.1 | XP_008762672.1 | 7 | 826 |