Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC292449 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY389467.1 | - | 1039 |
Gene Symbol: Atp13a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109369.1 | NP_001102839.1 | 5 | 635 |