Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCTN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281425.1 | NP_001268354.1 | 5, 6 | 440 | ||
NM_001281426.1 | NP_001268355.1 | 6, 7 | 555 | |||
NM_007234.4 | NP_009165.1 | 6, 7 | 524 | |||
NM_024348.3 | NP_077324.1 | 6, 7 | 621 | |||
GenBank mRNA | AF082513.1 | - | 6, 7 | 481 | ||
AK055199.1 | - | 1, 2 | 2696 | |||
AK091661.1 | - | 4, 5 | 3551 | |||
AK314730.1 | - | 6, 7 | 492 | |||
BC000319.2 | - | 6, 7 | 572 | 2821720 | ||
BC003004.1 | - | 6, 7 | 572 | 2821720 | ||
BC107697.1 | - | 6, 7 | 493 | 6454606 | ||
BG715167.1 | - | 6, 7 | 505 | |||
BI596981.1 | - | 6, 7 | 508 | |||
BI837796.1 | - | 5, 6 | 399 | |||
CN257020.1 | - | 4, 5 | 276 | |||
CR541889.1 | - | 6, 7 | 465 |