Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phemx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002728846.4 | XP_002728892.3 | 8 | 3078 | ||
XM_006230915.2 | XP_006230977.1 | 7 | 3003 | |||
XM_006230916.2 | XP_006230978.1 | 7 | 2991 | |||
GenBank mRNA | BC088266.1 | - | 5 | 369 | ||
BC158837.1 | - | 6 | 543 |