Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317192.1 | NP_001304121.1 | 4 | 410 | ||
NM_001317193.1 | NP_001304122.1 | 3 | 359 | |||
NM_001317194.1 | NP_001304123.1 | 5 | 421 | |||
NM_007241.3 | NP_009172.2 | 4 | 410 | |||
NR_133679.1 | 4 | 410 | ||||
XM_017024075.1 | XP_016879564.1 | 4 | 352 | |||
GenBank mRNA | AF156102.1 | - | 4 | 359 | ||
BC008976.1 | - | 4 | 360 | 3452153 | ||
BC038830.1 | - | 4 | 384 | 3900806 | ||
BE394098.1 | - | 5 | 323 | |||
BE818533.1 | - | 3 | 213 | |||
BG248541.1 | - | 4 | 399 | |||
BP342490.1 | - | 4 | 410 |