Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017414.3 | NP_059110.2 | 4, 5 | 741 | ||
XM_006724074.2 | XP_006724137.1 | 4, 5 | 812 | |||
GenBank mRNA | AF176642.2 | - | 3, 4 | 423 | ||
AJ243526.1 | - | 4, 5 | 606 | |||
AK313385.1 | - | 4, 5 | 562 | |||
AL136690.1 | - | 4, 5 | 562 | |||
BC014896.1 | - | 4, 5 | 547 | 3910720 | ||
BI752849.1 | - | 4, 5 | 636 | |||
BT006835.1 | - | 3, 4 | 403 | |||
CR457216.1 | - | 3, 4 | 403 | |||
CT841507.1 | - | 3, 4 | 469 |