Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDDM3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006683.4 | NP_006674.2 | 2 | 214 | ||
XM_017020934.1 | XP_016876423.1 | 2 | 231 | |||
GenBank mRNA | BC069407.1 | - | 2 | 185 | 7262138 | |
BC096698.1 | - | 2 | 204 | 40014618 | ||
BC096731.1 | - | 2 | 204 | 40014622 | ||
BC098164.1 | - | 1 | 204 | 40014621 | ||
BC098356.1 | - | 1 | 204 | 40014616 | ||
BC106711.1 | - | 1 | 168 | 40009240 | ||
BC106712.1 | - | 1 | 168 | 40009242 | ||
X76383.2 | - | 2 | 214 |