Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TLK2 SIRNA Assays ›
Gene Symbol:
TLK2
Gene Name:
tousled like kinase 2
Gene Aliases:
HsHPK, PKU-ALPHA
Chromosome Location:
Chr.17: 62477481 - 62615481 on Build GRCh38
UniGene ID:
Hs.445078
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s223176
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TLK2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001284333.1
NP_001271262.1
15
1552
NM_001284363.1
NP_001271292.1
12
1147
NM_006852.3
NP_006843.2
14
1489
XM_011524215.2
XP_011522517.1
13
1196
XM_011524216.2
XP_011522518.1
15
1316
XM_011524217.2
XP_011522519.1
15
1503
XM_011524218.2
XP_011522520.1
12
1130
XM_011524220.2
XP_011522522.1
12
1100
XM_011524221.2
XP_011522523.1
14
1214
XM_011524222.2
XP_011522524.1
15
1695
XM_011524223.2
XP_011522525.1
13
1357
XM_011524224.2
XP_011522526.1
13
1415
XM_011524225.2
XP_011522527.1
13
1363
XM_011524226.2
XP_011522528.1
15
1409
XM_011524227.2
XP_011522529.1
14
1463
XM_011524228.2
XP_011522530.1
15
1563
XM_011524229.2
XP_011522531.1
16
1490
XM_011524231.1
XP_011522533.1
14
1289
XM_017024044.1
XP_016879533.1
14
1323
XM_017024045.1
XP_016879534.1
13
1431
XM_017024046.1
XP_016879535.1
14
1253
XM_017024047.1
XP_016879536.1
14
1250
XM_017024048.1
XP_016879537.1
14
1711
XM_017024049.1
XP_016879538.1
14
1664
XM_017024050.1
XP_016879539.1
12
1196
XM_017024051.1
XP_016879540.1
13
1645
XM_017024052.1
XP_016879541.1
13
1596
XM_017024053.1
XP_016879542.1
13
1268
XM_017024054.1
XP_016879543.1
12
1294
XM_017024055.1
XP_016879544.1
11
1198
XM_017024056.1
XP_016879545.1
12
1349
XM_017024057.1
XP_016879546.1
12
1163
XM_017024058.1
XP_016879547.1
14
1441
XM_017024059.1
XP_016879548.1
14
1431
XM_017024060.1
XP_016879549.1
13
1348
GenBank mRNA
AB004884.1
-
12
1125
AF162667.1
-
14
1361
AK300586.1
-
12
1147
AX775745.1
-
14
1361
BC044925.1
-
15
1459
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
regulation of chromatin assembly or disassembly
protein phosphorylation
cellular response to DNA damage stimulus
cell cycle
chromosome segregation
negative regulation of autophagy
covalent chromatin modification
peptidyl-serine phosphorylation
negative regulation of proteasomal ubiquitin-dependent protein catabolic process
intracellular signal transduction
cellular response to gamma radiation
protein serine/threonine kinase activity
protein binding
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-7dvbj:80/10.239.167.51:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0