Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145047.1 | NP_001138519.1 | 12 | 836 | ||
NM_001145048.1 | NP_001138520.1 | 11 | 768 | |||
NM_001145049.1 | NP_001138521.1 | 9 | 661 | |||
NM_007266.3 | NP_009197.2 | 11 | 875 | |||
NR_026735.1 | 9 | 768 | ||||
GenBank mRNA | AB044661.1 | - | 11 | 836 | ||
AJ010842.1 | - | 11 | 765 | |||
AK298827.1 | - | 11 | 1169 | |||
AK298866.1 | - | 11 | 875 | |||
AK302131.1 | - | 9 | 734 | |||
AK310340.1 | - | 12 | 826 | |||
AK311423.1 | - | 9 | 791 | |||
AK312992.1 | - | 11 | 847 | |||
AK316404.1 | - | 9 | 661 | |||
BC007451.2 | - | 11 | 819 | 3029376 |