Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CKB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001823.4 | NP_001814.2 | 7 | 916 | ||
XM_017020951.1 | XP_016876440.1 | 6 | 962 | |||
XM_017020952.1 | XP_016876441.1 | 6 | 766 | |||
GenBank mRNA | AK290101.1 | - | 7 | 887 | ||
AK298198.1 | - | 7 | 785 | |||
AK312282.1 | - | 7 | 888 | |||
BC001190.1 | - | 7 | 875 | 3354679 | ||
BC004914.2 | - | 7 | 867 | 3537262 | ||
BC008323.1 | - | 7 | 873 | 3358085 | ||
BC010002.2 | - | 7 | 882 | 4135106 | ||
BC019259.1 | - | 7 | 871 | 3503057 | ||
BC019281.1 | - | 7 | 881 | 2823530 | ||
CR542268.1 | - | 6 | 810 | |||
EU794629.1 | - | 7 | 871 | |||
FJ224337.1 | - | 6 | 810 | |||
KU177977.1 | - | 6 | 810 | |||
KU177978.1 | - | 5 | 882 | |||
L47647.1 | - | 7 | 853 | |||
M16364.1 | - | 6 | 818 | |||
M16451.1 | - | 6 | 833 |