Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127198.2 | NP_001120670.1 | 13 | 1917 | ||
NM_001321185.1 | NP_001308114.1 | 13 | 1870 | |||
NM_007267.7 | NP_009198.4 | 13 | 1821 | |||
XM_005256995.1 | XP_005257052.1 | 13 | 1946 | |||
XM_011524255.1 | XP_011522557.1 | 12 | 1848 | |||
XM_011524256.1 | XP_011522558.1 | 12 | 1722 | |||
XM_011524257.2 | XP_011522559.1 | 7 | 966 | |||
XM_017024109.1 | XP_016879598.1 | 11 | 1637 | |||
GenBank mRNA | AB002405.2 | - | 5 | 629 | ||
AK021738.1 | - | 10 | 1858 | |||
AK074065.1 | - | 13 | 1820 | |||
AK096076.1 | - | 13 | 1853 | |||
AK310734.1 | - | 12 | 2241 | |||
AY057379.1 | - | 12 | 1660 | |||
AY099356.1 | - | 14 | 1819 | |||
BC007766.1 | - | 12 | 1560 | |||
BC018346.1 | - | 6 | 785 | |||
BC023597.2 | - | 13 | 1801 | 4566166 | ||
BC035648.1 | - | 6 | 841 | 5575949 |