Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STARD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308056.1 | NP_001294985.1 | 6 | 700 | ||
NM_001308059.1 | NP_001294988.1 | 7 | 735 | |||
NM_001308060.1 | NP_001294989.1 | 4 | 465 | |||
NM_001308061.1 | NP_001294990.1 | 5 | 592 | |||
NM_139164.2 | NP_631903.1 | 6 | 706 | |||
NR_131754.1 | 5 | 489 | ||||
XM_017009041.1 | XP_016864530.1 | 5 | 517 | |||
XM_017009042.1 | XP_016864531.1 | 5 | 511 | |||
XM_017009043.1 | XP_016864532.1 | 4 | 429 | |||
XM_017009044.1 | XP_016864533.1 | 5 | 499 | |||
GenBank mRNA | AF480299.1 | - | 5 | 484 | ||
AK054566.1 | - | 6 | 568 | |||
AK125317.1 | - | 5 | 427 | |||
AK290312.1 | - | 6 | 619 | |||
AK315863.1 | - | 4 | 403 | |||
AK315869.1 | - | 5 | 517 | |||
AL710253.1 | - | 6 | 665 | |||
AL832599.2 | - | 2 | 162 |