Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH1L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034173.3 | NP_001029345.2 | 20 | 2475 | ||
NR_027752.1 | 20 | 2427 | ||||
XM_011537986.2 | XP_011536288.1 | 18 | 2034 | |||
XM_017018889.1 | XP_016874378.1 | 21 | 2681 | |||
GenBank mRNA | AK300373.1 | - | 20 | 2370 | ||
BC103934.1 | - | 20 | 2370 | 40008174 | ||
BC103935.1 | - | 20 | 2322 | 40008176 | ||
CR627287.1 | - | 16 | 2339 | |||
CR749561.1 | - | 1 | 67 |