Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305102.1 | NP_001292031.1 | 14 | 1625 | ||
NM_007271.3 | NP_009202.1 | 14 | 1628 | |||
XM_006714988.3 | XP_006715051.1 | 14 | 2008 | |||
XM_006714989.3 | XP_006715052.1 | 14 | 2005 | |||
GenBank mRNA | AK312559.1 | - | 14 | 1536 | ||
BC012085.1 | - | 14 | 1582 | 4554884 | ||
BC095413.1 | - | 14 | 1588 | 30337276 | ||
Z35102.1 | - | 13 | 1891 |