Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLPP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030059.2 | NP_001025230.1 | 5 | 722 | ||
XM_005269592.2 | XP_005269649.1 | 5 | 722 | |||
XM_006717685.3 | XP_006717748.1 | 5 | 391 | |||
XM_006717686.3 | XP_006717749.1 | 5 | 722 | |||
XM_011539444.2 | XP_011537746.1 | 6 | 449 | |||
XM_011539445.2 | XP_011537747.1 | 5 | 722 | |||
XM_017015820.1 | XP_016871309.1 | 5 | 394 | |||
XM_017015821.1 | XP_016871310.1 | 2 | 132 | |||
XM_017015822.1 | XP_016871311.1 | 2 | 99 | |||
GenBank mRNA | AK098668.1 | - | 5 | 305 | ||
AK296273.1 | - | 5 | 429 | |||
AK310178.1 | - | 5 | 373 | |||
BC132787.1 | - | 5 | 545 | 40146809 | ||
BI550027.1 | - | 5 | 454 | |||
CR739184.1 | - | 4 | 335 |