Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OTP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032109.2 | NP_115485.1 | 2 | 545 | ||
GenBank mRNA | BC001800.1 | - | 2 | 545 | 3354882 |