Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324444.1 | NP_001311373.1 | 6 | 599 | ||
NM_001324445.1 | NP_001311374.1 | 5 | 1314 | |||
NM_001324446.1 | NP_001311375.1 | 6 | 1332 | |||
NM_001324448.1 | NP_001311377.1 | 6 | 541 | |||
NM_001324449.1 | NP_001311378.1 | 5 | 1314 | |||
NM_001324450.1 | NP_001311379.1 | 6 | 599 | |||
NM_001324451.1 | NP_001311380.1 | 5 | 1372 | |||
NM_001324452.1 | NP_001311381.1 | 4 | 1259 | |||
NM_001324453.1 | NP_001311382.1 | 4 | 1259 | |||
NM_012091.4 | NP_036223.2 | 6 | 541 | |||
GenBank mRNA | AF125188.1 | - | 5 | 421 | ||
AK001687.1 | - | 6 | 532 | |||
AK296834.1 | - | 7 | 412 | |||
AK314652.1 | - | 6 | 483 | |||
BC002758.2 | - | 6 | 487 | 3632079 | ||
DC395938.1 | - | 6 | 541 |