Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABARAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007278.1 | NP_009209.1 | 1 | 168 | ||
GenBank mRNA | AB030711.1 | - | 1 | 167 | ||
AF044671.1 | - | 1 | 142 | |||
AF067171.1 | - | 1 | 168 | |||
AF161586.1 | - | 1 | 132 | |||
AF183425.1 | - | 1 | 167 | |||
AK098634.1 | - | 1 | 172 | |||
AK312205.1 | - | 1 | 163 | |||
BC106748.1 | - | 1 | 110 | 40022383 | ||
BC106749.1 | - | 1 | 110 | 40022384 | ||
CR457043.1 | - | 1 | 64 | |||
CR542235.1 | - | 1 | 64 | |||
JN663880.1 | - | 1 | 69 |