Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VAX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012476.2 | NP_036608.1 | 3 | 924 | ||
GenBank mRNA | AB593145.1 | - | 3 | 933 | ||
BC006336.2 | - | 3 | 899 | 4304066 | ||
Y17791.1 | - | 3 | 924 |