Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLA2G2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271814.1 | NP_001258743.1 | 3 | 1102 | ||
NM_012400.3 | NP_036532.1 | 4 | 1209 | |||
GenBank mRNA | AF188625.1 | - | 4 | 1176 | ||
AK026347.1 | - | 1 | 319 | |||
BC025706.1 | - | 4 | 1158 | 5223912 |