Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014372.4 | NP_055187.1 | 3 | 852 | ||
GenBank mRNA | AB024703.1 | - | 3 | 495 | ||
AF151881.1 | - | 3 | 495 | |||
AK313140.1 | - | 3 | 614 | |||
BC020964.2 | - | 3 | 447 | 3917224 | ||
BC047654.1 | - | 3 | 820 | 5261966 |