Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098202.1 | NP_001091672.1 | 2 | 1300 | ||
NM_006497.3 | NP_006488.2 | 2 | 1403 |