Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006638.2 | NP_001006639.1 | 6, 7 | 499 | ||
NM_001163989.1 | NP_001157461.1 | 6, 7 | 648 | |||
NM_001163990.1 | NP_001157462.1 | 5, 6 | 388 | |||
NM_175738.4 | NP_783865.1 | 6, 7 | 892 | |||
XM_005257288.4 | XP_005257345.1 | 6, 7 | 517 | |||
GenBank mRNA | AK054846.1 | - | 6 | 1328 | ||
AK057069.1 | - | 1 | 43 | |||
AK098068.1 | - | 6, 7 | 499 | |||
AK127073.1 | - | 6, 7 | 1431 | |||
AK290202.1 | - | 6, 7 | 868 | |||
AK296062.1 | - | 6, 7 | 487 | |||
AK296172.1 | - | 5, 6 | 351 | |||
AK298033.1 | - | 5 | 883 | |||
AK303442.1 | - | 6, 7 | 551 | |||
BC016615.1 | - | 6, 7 | 445 | 4520191 | ||
BC040547.1 | - | 6, 7 | 878 | 5287038 |