Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007359.4 | NP_031385.2 | 5 | 838 | ||
XM_005257163.1 | XP_005257220.1 | 5 | 616 | |||
GenBank mRNA | AK292425.1 | - | 5 | 777 | ||
AK296682.1 | - | 5 | 779 | |||
AK310635.1 | - | 5 | 784 | |||
BC030008.1 | - | 5 | 573 | |||
BC044656.1 | - | 5 | 721 | 5547561 | ||
BC050526.1 | - | 5 | 741 | 5561860 | ||
X80199.2 | - | 5 | 776 |