Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000207.2 | NP_000198.1 | 3 | 325 | ||
NM_001185097.1 | NP_001172026.1 | 3 | 351 | |||
NM_001185098.1 | NP_001172027.1 | 2 | 504 | |||
NM_001291897.1 | NP_001278826.1 | 3 | 385 | |||
GenBank mRNA | AY899304.1 | - | 3 | 321 | ||
BC005255.1 | - | 3 | 325 | 3950204 | ||
BM510347.1 | - | 3 | 346 | |||
BM510748.1 | - | 3 | 329 | |||
BP322143.1 | - | 2 | 504 | |||
BT006808.1 | - | 2 | 266 | |||
DQ778082.1 | - | 2 | 230 | |||
JF909299.1 | - | 2 | 219 | |||
JQ951950.1 | - | 3 | 310 | |||
X70508.1 | - | 3 | 310 |