Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NID2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007361.3 | NP_031387.3 | 5 | 1322 | ||
XM_005267405.4 | XP_005267462.1 | 4 | 1406 | |||
XM_005267406.4 | XP_005267463.1 | 4 | 1406 | |||
XM_005267407.4 | XP_005267464.1 | 5 | 1325 | |||
GenBank mRNA | AJ223500.1 | - | 5 | 1088 | ||
AK291652.1 | - | 5 | 1150 | |||
AK300462.1 | - | 6 | 960 | |||
AL701794.1 | - | 4 | 475 | |||
BX648241.1 | - | 6 | 1095 | |||
D86425.1 | - | 5 | 1091 |