Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAMB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002292.3 | NP_002283.3 | 25 | 4219 | ||
XM_005265127.3 | XP_005265184.1 | 26 | 4018 | |||
GenBank mRNA | AK094050.1 | - | 6 | 838 | ||
S77512.1 | - | 25 | 4072 | |||
X72760.1 | - | 2 | 374 | |||
X79683.1 | - | 26 | 4085 |