Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCN9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001676.1 | NP_001001676.1 | 2 | 146 | ||
XM_017014712.1 | XP_016870201.1 | 2 | 146 | |||
GenBank mRNA | AY301270.1 | - | 2 | 146 | ||
EU794679.1 | - | 2 | 146 |