Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MXI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008541.1 | NP_001008541.1 | 4, 5 | 506 | ||
NM_005962.4 | NP_005953.4 | 5, 6 | 731 | |||
NM_130439.3 | NP_569157.2 | 5, 6 | 929 | |||
GenBank mRNA | AK303949.1 | - | 5, 6 | 608 | ||
AK307090.1 | - | 6, 7 | 617 | |||
AK309718.1 | - | 5, 6 | 544 | |||
AL538051.3 | - | 5, 6 | 742 | |||
BC012907.1 | - | 4, 5 | 506 | 3882557 | ||
BC035128.1 | - | 5, 6 | 935 | 5263647 | ||
BT007069.1 | - | 3, 4 | 386 | |||
BX417051.2 | - | 5, 6 | 727 | |||
CR536576.1 | - | 5, 6 | 524 | |||
D63940.1 | - | 4, 5 | 416 | |||
KU178207.1 | - | 3, 4 | 386 | |||
KU178208.1 | - | 4, 5 | 432 | |||
KU178209.1 | - | 4, 5 | 416 |