Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199397.1 | NP_001186326.1 | 6 | 930 | ||
NM_001199398.1 | NP_001186327.1 | 6 | 930 | |||
NM_001199399.1 | NP_001186328.1 | 6 | 930 | |||
NM_001199400.1 | NP_001186329.1 | 5 | 1008 | |||
NM_012224.2 | NP_036356.1 | 5 | 1008 | |||
XM_006714228.1 | XP_006714291.1 | 6 | 932 | |||
XM_011532003.1 | XP_011530305.1 | 6 | 932 | |||
XM_011532004.1 | XP_011530306.1 | 6 | 932 | |||
XM_011532005.2 | XP_011530307.1 | 6 | 932 | |||
XM_017008249.1 | XP_016863738.1 | 5 | 834 | |||
XM_017008250.1 | XP_016863739.1 | 5 | 1007 | |||
XM_017008251.1 | XP_016863740.1 | 5 | 834 | |||
XM_017008252.1 | XP_016863741.1 | 4 | 909 | |||
XM_017008253.1 | XP_016863742.1 | 6 | 932 | |||
GenBank mRNA | AB067488.1 | - | 5 | 844 | ||
AK027580.1 | - | 2 | 84 | |||
AL050385.1 | - | 6 | 885 | |||
BC015147.1 | - | 5 | 770 | |||
BC068201.1 | - | 6 | 890 | |||
BC114491.1 | - | 6 | 690 | 40082305 | ||
CR933642.1 | - | 6 | 885 | |||
CR936672.1 | - | 5 | 965 |