Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLCA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012128.3 | NP_036260.2 | 7 | 1090 | ||
NR_024602.1 | 6 | 981 | ||||
XM_011541015.2 | XP_011539317.1 | 7 | 909 | |||
GenBank mRNA | AF127035.1 | - | 7 | 1074 | ||
AK000072.1 | - | 7 | 1088 | |||
AK293766.1 | - | 6 | 745 | |||
AK307785.1 | - | 6 | 926 | |||
AK307898.1 | - | 5 | 695 | |||
AK314430.1 | - | 7 | 1086 | |||
AY358470.1 | - | 7 | 1070 | |||
BC113687.1 | - | 7 | 1085 | 8322739 | ||
BC113689.1 | - | 7 | 1084 | 8322741 |