Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321732.1 | NP_001308661.1 | 7 | 1319 | ||
NM_001321830.1 | NP_001308759.1 | 8 | 1111 | |||
NM_016222.3 | NP_057306.2 | 8 | 1042 | |||
GenBank mRNA | AF195417.1 | - | 8 | 659 | ||
AK001255.1 | - | 8 | 667 | |||
AK027768.1 | - | 8 | 677 | |||
AK091545.1 | - | 6 | 1889 | |||
AK091774.1 | - | 7 | 954 | |||
AK092585.1 | - | 5 | 1329 | |||
AK222598.1 | - | 8 | 667 | |||
AK315491.1 | - | 8 | 677 | |||
BC015476.1 | - | 8 | 682 | 3917178 | ||
BX641072.1 | - | 6 | 726 |