Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other IKZF2 SIRNA Assays ›
Gene Symbol:
IKZF2
Gene Name:
IKAROS family zinc finger 2
Gene Aliases:
ANF1A2, HELIOS, ZNF1A2, ZNFN1A2
Chromosome Location:
Chr.2: 212999684 - 213152753 on Build GRCh38
UniGene ID:
Hs.604950
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s22420
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: IKZF2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001079526.1
NP_001072994.1
9
1355
NM_016260.2
NP_057344.2
8
1286
XM_005246384.4
XP_005246441.1
8
1396
XM_005246385.3
XP_005246442.1
8
1426
XM_005246386.4
XP_005246443.1
8
1277
XM_011510802.2
XP_011509104.1
9
1468
XM_011510803.2
XP_011509105.1
9
1317
XM_011510804.2
XP_011509106.1
9
1240
XM_011510805.2
XP_011509107.1
9
1329
XM_011510807.2
XP_011509109.1
9
1504
XM_011510808.2
XP_011509110.1
9
1354
XM_011510809.2
XP_011509111.1
9
1366
XM_011510810.2
XP_011509112.1
9
1280
XM_011510811.2
XP_011509113.1
8
1401
XM_011510812.2
XP_011509114.1
8
1262
XM_011510814.2
XP_011509116.1
8
1760
XM_011510815.2
XP_011509117.1
8
1208
XM_011510816.2
XP_011509118.1
6
1209
XM_011510817.2
XP_011509119.1
7
1144
XM_011510818.2
XP_011509120.1
6
6863
XM_011510819.2
XP_011509121.1
7
1133
XM_017003589.1
XP_016859078.1
8
1348
XM_017003590.1
XP_016859079.1
8
1318
XM_017003591.1
XP_016859080.1
6
1751
XM_017003592.1
XP_016859081.1
7
1065
GenBank mRNA
AF130863.1
-
8
1286
AK301503.1
-
8
1282
AY008294.1
-
2
1750
AY587062.1
-
9
1270
AY587063.1
-
9
1232
AY587064.1
-
9
1316
AY587065.1
-
8
1123
AY587066.1
-
6
895
AY587067.1
-
7
934
AY587068.1
-
8
1194
AY587069.1
-
7
1039
AY587070.1
-
5
682
AY587071.1
-
6
901
AY587072.1
-
3
229
AY587073.1
-
8
1296
BC028936.1
-
9
1355
4540576
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
biological_process
positive regulation of transcription from RNA polymerase II promoter
molecular_function
transcription factor activity, sequence-specific DNA binding
protein binding
protein homodimerization activity
transcription regulatory region DNA binding
metal ion binding
protein heterodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0