Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164391.1 | NP_001157863.1 | 14 | 1519 | ||
NM_001164392.1 | NP_001157864.1 | 12 | 1428 | |||
NM_001164393.1 | NP_001157865.1 | 11 | 1318 | |||
NM_007358.3 | NP_031384.1 | 13 | 1599 | |||
XM_011541016.1 | XP_011539318.1 | 13 | 1555 | |||
XM_011541019.1 | XP_011539321.1 | 13 | 1347 | |||
XM_017000676.1 | XP_016856165.1 | 13 | 1454 | |||
XM_017000677.1 | XP_016856166.1 | 12 | 1384 | |||
XM_017000678.1 | XP_016856167.1 | 13 | 1337 | |||
XM_017000679.1 | XP_016856168.1 | 12 | 1431 | |||
GenBank mRNA | AF072814.1 | - | 12 | 1297 | ||
AF073293.1 | - | 3 | 801 | |||
AF086908.1 | - | 12 | 1400 | |||
AJ010014.1 | - | 13 | 1550 | |||
AK290318.1 | - | 13 | 1551 | |||
AK302776.1 | - | 14 | 1457 | |||
AK302900.1 | - | 11 | 1270 | |||
BC010013.2 | - | 12 | 1352 | 4308565 |