Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LTB4R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164692.2 | NP_001158164.1 | 2 | 1013 | ||
NM_019839.4 | NP_062813.2 | 2 | 957 | |||
GenBank mRNA | AB029892.1 | - | 1 | 842 | ||
AB032558.1 | - | 2 | 923 | |||
AB044402.1 | - | 1 | 842 | |||
AF277230.1 | - | 1 | 2553 | |||
AF308571.1 | - | 1 | 842 | |||
BC038437.1 | - | 2 | 993 |
Set Membership: |
G-protein coupled receptor |