Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRIP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020645.2 | NP_065696.1 | 6 | 742 | ||
GenBank mRNA | AK024750.1 | - | 6 | 695 | ||
AK098694.1 | - | 6 | 695 | |||
AK296484.1 | - | 5 | 554 | |||
AY211930.1 | - | 5 | 439 | |||
BC008835.2 | - | 6 | 678 | 4304488 |