Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRNA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303034.1 | NP_001289963.1 | 5 | 1079 | ||
NM_001303035.1 | NP_001289964.1 | 5 | 1012 | |||
NM_020402.3 | NP_065135.2 | 5 | 1017 | |||
XM_011520234.1 | XP_011518536.1 | 4 | 4213 | |||
GenBank mRNA | AF199235.2 | - | 5 | 1017 | ||
AJ278118.1 | - | 5 | 987 | |||
AJ295237.1 | - | 5 | 987 | |||
BC136250.1 | - | 5 | 995 | |||
BC143303.1 | - | 5 | 1062 |