Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLGN4Y | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206850.1 | NP_001193779.1 | 6 | 1568 | ||
NM_014893.4 | NP_055708.3 | 6 | 1898 | |||
NR_028319.1 | 6 | 2120 | ||||
NR_046355.1 | 7 | 1885 | ||||
XM_006724874.2 | XP_006724937.1 | 7 | 2130 | |||
XM_011531424.2 | XP_011529726.1 | 7 | 2448 | |||
XM_011531425.2 | XP_011529727.1 | 7 | 2220 | |||
XM_011531426.2 | XP_011529728.1 | 7 | 2181 | |||
XM_011531427.1 | XP_011529729.1 | 7 | 1935 | |||
XM_011531428.1 | XP_011529730.1 | 8 | 2116 | |||
XM_011531429.2 | XP_011529731.1 | 7 | 2731 | |||
XM_011531430.2 | XP_011529732.1 | 7 | 2069 | |||
XM_017030034.1 | XP_016885523.1 | 6 | 2160 | |||
XM_017030035.1 | XP_016885524.1 | 6 | 2081 | |||
XM_017030036.1 | XP_016885525.1 | 6 | 1875 | |||
XM_017030037.1 | XP_016885526.1 | 6 | 1873 | |||
XM_017030038.1 | XP_016885527.1 | 6 | 2671 | |||
XM_017030039.1 | XP_016885528.1 | 7 | 1406 | |||
XM_017030040.1 | XP_016885529.1 | 6 | 1255 | |||
GenBank mRNA | AB023168.1 | - | 6 | 1424 | ||
AF376804.1 | - | 6 | 1898 | |||
AK295117.1 | - | 7 | 1885 | |||
BC113525.1 | - | 5 | 1725 | 8322577 | ||
BC113551.1 | - | 5 | 1725 | 8322603 | ||
BX537428.1 | - | 6 | 2121 |