Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPCS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021928.3 | NP_068747.1 | 4 | 469 | ||
GenBank mRNA | AK026302.1 | - | 4 | 444 | ||
AK092634.1 | - | 3 | 1452 | |||
AL136660.1 | - | 4 | 439 | |||
AY359044.1 | - | 4 | 427 | |||
BC047290.1 | - | 4 | 412 | 5224927 | ||
BC053883.1 | - | 3 | 2363 | |||
DB285025.1 | - | 4 | 471 |