Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROCK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005406.2 | NP_005397.1 | 16, 17 | 2827 | ||
XM_011526137.2 | XP_011524439.1 | 5, 6 | 1061 | |||
GenBank mRNA | AB208965.1 | - | 8, 9 | 920 | ||
AK057038.1 | - | 16, 17 | 2828 | |||
AK307388.1 | - | 16, 17 | 2642 | |||
BC113114.1 | - | 16, 17 | 1902 | 7961989 | ||
U43195.1 | - | 16, 17 | 1886 |