Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001247999.1 | NP_001234928.1 | 4 | 732 | ||
NM_022349.3 | NP_071744.2 | 3 | 543 | |||
NM_152851.2 | NP_690590.1 | 3 | 543 | |||
NM_152852.2 | NP_690591.1 | 4 | 668 | |||
XM_005274177.2 | XP_005274234.1 | 4 | 727 | |||
XM_006718658.2 | XP_006718721.1 | 4 | 602 | |||
XM_011545209.1 | XP_011543511.1 | 4 | 602 | |||
XM_017018125.1 | XP_016873614.1 | 4 | 602 | |||
GenBank mRNA | AB013104.1 | - | 3 | 251 | ||
AF142409.1 | - | 3 | 271 | |||
AF172819.1 | - | 4 | 689 | |||
AF212240.1 | - | 3 | 412 | |||
AF237908.1 | - | 3 | 413 | |||
AF253977.1 | - | 3 | 413 | |||
AF261136.1 | - | 3 | 410 | |||
AF286866.1 | - | 2 | 191 | |||
AF350502.1 | - | 3 | 260 | |||
AF350503.1 | - | 3 | 260 | |||
AF354930.1 | - | 3 | 413 | |||
AK026454.1 | - | 2 | 1981 | |||
AK291870.1 | - | 4 | 668 | |||
AK300725.1 | - | 3 | 277 | |||
BC022854.1 | - | 3 | 282 | 3934920 | ||
BC108728.1 | - | 3 | 316 | 6196213 | ||
DA917243.1 | - | 3 | 367 | |||
DA979138.1 | - | 3 | 314 |