Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRPK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003137.4 | NP_003128.3 | 15 | 1893 | ||
NR_034069.1 | 15 | 1921 | ||||
GenBank mRNA | AJ318054.1 | - | 14 | 2291 | ||
AK292277.1 | - | 15 | 1838 | |||
AK299591.1 | - | 15 | 1881 | |||
AK312712.1 | - | 15 | 1785 | |||
BC038292.1 | - | 15 | 1858 | 4824261 | ||
BX648663.1 | - | 15 | 1810 | |||
BX648697.1 | - | 15 | 1811 |