Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014901.4 | NP_055716.1 | 3 | 802 | ||
XM_005265840.2 | XP_005265897.1 | 3 | 514 | |||
XM_005265841.4 | XP_005265898.1 | 3 | 759 | |||
XM_005265842.4 | XP_005265899.1 | 3 | 744 | |||
XM_005265843.2 | XP_005265900.1 | 3 | 450 | |||
XM_005265844.3 | XP_005265901.1 | 3 | 555 | |||
XM_005265845.4 | XP_005265902.1 | 3 | 693 | |||
XM_006714831.2 | XP_006714894.1 | 3 | 514 | |||
XM_006714832.2 | XP_006714895.1 | 3 | 514 | |||
XM_011534466.2 | XP_011532768.1 | 3 | 517 | |||
XM_011534467.2 | XP_011532769.1 | 3 | 748 | |||
GenBank mRNA | AB029023.1 | - | 3 | 687 | ||
AK302385.1 | - | 3 | 555 | |||
BC039833.1 | - | 3 | 758 | 5535911 | ||
BC063297.1 | - | 3 | 803 | 30349418 |