Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAPPC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003274.4 | NP_003265.3 | 5 | 733 | ||
XM_005261168.4 | XP_005261225.1 | 5 | 726 | |||
XM_011529714.2 | XP_011528016.1 | 5 | 726 | |||
XM_011529715.2 | XP_011528017.1 | 5 | 726 | |||
XM_011529716.2 | XP_011528018.1 | 5 | 726 | |||
XM_011529717.2 | XP_011528019.1 | 5 | 726 | |||
XM_011529718.2 | XP_011528020.1 | 5 | 726 | |||
XM_011529720.2 | XP_011528022.1 | 5 | 726 | |||
XM_011529725.2 | XP_011528027.1 | 5 | 726 | |||
XM_011529726.2 | XP_011528028.1 | 5 | 726 | |||
XM_011529728.2 | XP_011528030.1 | 2 | 243 | |||
XM_011529729.2 | XP_011528031.1 | 2 | 120 | |||
XM_011529730.2 | XP_011528032.1 | 5 | 726 | |||
XM_017028454.1 | XP_016883943.1 | 5 | 726 | |||
GenBank mRNA | BC046241.1 | - | 5 | 711 | ||
BC052247.1 | - | 5 | 711 | |||
BC094823.1 | - | 5 | 711 | |||
BC101728.1 | - | 5 | 653 | 8069234 | ||
BX370317.2 | - | 2 | 213 | |||
U19252.1 | - | 5 | 701 | |||
U61500.1 | - | 5 | 717 |