Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003285.2 | NP_003276.3 | 14 | 3336 | ||
XM_011509949.2 | XP_011508251.1 | 13 | 2945 | |||
XM_017002218.1 | XP_016857707.1 | 15 | 3773 | |||
XM_017002219.1 | XP_016857708.1 | 14 | 3701 | |||
GenBank mRNA | BC129830.1 | - | 12 | 2659 | 40114168 | |
X98085.1 | - | 13 | 2898 | |||
Z67996.1 | - | 12 | 2862 |